missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F6 Recombinant Rabbit Monoclonal Antibody (23GB5255), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | E2F6 |
|---|---|
| Clone | 23GB5255 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088419
![]() |
Invitrogen™
MA552824 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087416
|
Invitrogen™
MA552825 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E2F6 Recombinant Monoclonal antibody specifically detects E2F6 in Human samples. It is validated for Western BlotSpecifications
| E2F6 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75461 | |
| 1876 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5255 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI462434; E2F transcription factor 6; E2f6; E2F-6; E2F6b; E2F-binding site-modulating activity protein; EMA; MGC111545; Transcription factor E2F6; transcriptional repressor E2F6 | |
| A synthesized peptide derived from human E2F6 (100-281AA). | |
| Primary | |
| -20°C | |
| E2F6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title