Learn More
Invitrogen™ E2F5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585578
Description
Keep as concentrated solution. Predicted reactivity: Mouse (91%). Positive Control: A549, H1299, HCT116, mouse placenta, HA-human E2F5-transfected 293T, mouse leukocyte treated with CFA. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E2F5 is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionarily conserved domains that are present in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein is differentially phosphorylated and is expressed in a wide variety of human tissues. It has higher identity to E2F4 than to other family members. Both this protein and E2F4 interact with tumor suppressor proteins p130 and p107, but not with pRB. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
| E2F5 | |
| Polyclonal | |
| Unconjugated | |
| E2F5 | |
| AU024671; E2F transcription factor 5; E2F transcription factor 5, p130-binding; E2f5; E2F-5; Transcription factor E2F5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13559, 1875 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q15329, Q61502 | |
| E2F5 | |
| Recombinant protein encompassing a sequence within the center region of human E2F5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.