missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56500
This item is not returnable.
View return policy
Description
E2F5 Polyclonal specifically detects E2F5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| E2F5 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| E2F transcription factor 5, p130-binding, E2F-5, transcription factor E2F5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| E2F5 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:INLKSHSGPIHVLLINKESSSSKPVVFPVPPPDDLTQPSSQSLTPVTPQKSSMATQNL | |
| 100 μL | |
| Cell Cycle and Replication | |
| 1875 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction