missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F4 Recombinant Rabbit Monoclonal Antibody (24GB5375)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557392
This item is not returnable.
View return policy
Description
E2F4 Recombinant Monoclonal Antibody for Western Blot, Flow
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
Specifications
| E2F4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| E2f4 | |
| 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1874 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5375 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16254 | |
| E2f4 | |
| A synthesized peptide derived from human E2F4 (aa 286-299). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction