missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579182
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, U20S whole cell, MCF-7 whole cell. IHC: mouse intestine tissue, rat intestine tissue, human intetsinal cancer tissue.
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
Specifications
| E2F4 | |
| Polyclonal | |
| Unconjugated | |
| E2f4 | |
| 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100360427, 104394, 1874 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| Q16254, Q8R0K9 | |
| E2f4 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human E2F4 (106-144aa ELQQREQELDQHKVWVQQSIRNVTEDVQNSCLAYVTHED). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction