missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F4 Monoclonal Antibody (4E2F04 (WUF10)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511276
This item is not returnable.
View return policy
Description
MA5-11276 targets E2F-4 Transcription Factor in GS, IF, IHC (P), and WB applications and shows reactivity with Human and Rat samples. The MA5-11276 immunogen is recombinant human E2F-4 protein.
E2F4 Monoclonal antibody specifically detects E2F4 in Human, Mouse, Rat samples. It is validated for Gel Shift, Immunohistochemistry (Paraffin), Western Blot.
Specifications
| E2F4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Q16254, Q8R0K9 | |
| E2f4 | |
| Recombinant human E2F-4 protein | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Gel Shift, Immunohistochemistry (Paraffin), Western Blot | |
| 4E2F04 (WUF10) | |
| Unconjugated | |
| E2f4 | |
| 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100360427, 104394, 1874 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction