missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F4/E2F5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536051
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 44 kDa. Purity is >95% by SDS-PAGE.
E2F was originally identified through its role in transcriptional activation of the adenovirus E2 promoter. Sequences homologous to the E2F binding site have been found upstream of a number of genes that encode proteins with putative functions in the G1 and S phases of the cell cycle. E2F-1 is a member of a broader family of transcriptional regulators including E2F-2, E2F-3, E2F-4, E2F-5 and E2F-6, each of which forms heterodimers with a second protein, DP-1, forming an active E2F transcriptional regulatory complex.
Specifications
| E2F4/E2F5 | |
| Polyclonal | |
| Unconjugated | |
| E2F4 | |
| 2010111M04Rik; AI427446; AU024671; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F transcription factor 5; E2F transcription factor 5, p130-binding; E2F4; E2F-4; E2f5; E2F-5; p107/p130-binding protein; transcription factor E2F4; Transcription factor E2F5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100360427, 104394, 116651, 13559, 1874, 1875 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15329, Q16254, Q61502, Q62814, Q8R0K9 | |
| E2F4, E2F5 | |
| Synthetic peptide corresponding to amino acids 18-72 of human E2F4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction