missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584678
This item is not returnable.
View return policy
Description
Immunogen sequence: TNNQDHNGNI PKPASKDLAS TNSGHSDCSV SMGNLSPLAS PANLLQQTED QIPSNLEGPF VNLLPPLLQE DYLL Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F1 and E2F2, have an additional cyclin binding domain. This protein binds specifically to retinoblastoma protein pRB in a cell-cycle dependent manner.
Specifications
| E2F3 | |
| Polyclonal | |
| Unconjugated | |
| E2F3 | |
| DKFZp686C18211; E2F transcription factor 3; E2f3; E2F-3; E2f3a; E2F3b; KIAA0075; MGC104598; RGD1561600; RP1-177P22.2; Transcription factor E2F3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1871 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00716 | |
| E2F3 | |
| Recombinant protein corresponding to Human E2F3. Recombinant protein control fragment (Product #RP-107394). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction