missing translation for 'onlineSavingsMsg'
Learn More

E2F3 Antibody (3C11), Novus Biologicals™

Product Code. 18338497 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338497 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338497 Supplier Novus Biologicals Supplier No. H00001871M04100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

E2F3 Monoclonal antibody specifically detects E2F3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3C11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH16847.1
Gene Alias DKFZp686C18211, E2F transcription factor 3, E2F-3, KIAA0075, MGC104598, transcription factor E2F3
Host Species Mouse
Immunogen E2F3 (AAH16847.1, 1 a.a. ~ 133 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MQSGGGVKTDDTSTLNSLCGYAWVYVWEEKQRCRLSSFFSSSASIPGLLPSHTLDLVQNVGVVLDEALGWGRERELCVKCLLEMHCGVFSCMGNHLCQAFPHFPYLSHLVSCLCFQLCVILFASCTKLIFSKV
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1871
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.