missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F1 Recombinant Rabbit Monoclonal Antibody (23GB1505), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA553965
This item is not returnable.
View return policy
Description
E2F1 Recombinant Monoclonal antibody specifically detects E2F1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| E2F1 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O09139, Q01094, Q61501 | |
| E2F1 | |
| A synthesized peptide derived from human E2F1 (355-400AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1505 | |
| Unconjugated | |
| E2F1 | |
| E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13555, 1869, 399489 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction