missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F1 Monoclonal Antibody (KH95)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 321400
This item is not returnable.
View return policy
Description
E2F1 Monoclonal Antibody for Western Blot, IHC, IP, ChIP
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F2 and E2F3, have an additional cyclin binding domain. This protein binds preferentially to retinoblastoma protein pRB in a cell-cycle dependent manner. It can mediate both cell proliferation and p53-dependent/independent apoptosis.
Specifications
| E2F1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| O09139, Q01094, Q61501 | |
| E2F1 | |
| Recombinant human E2F-1 protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| KH95 | |
| Unconjugated | |
| E2F1 | |
| E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 13555, 1869, 399489 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction