Learn More
Invitrogen™ E2F1 Monoclonal Antibody (8G9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517070
Description
MA5-17070 targets E2F1 in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human and Mouse samples. The MA5-17070 immunogen is purified recombinant fragment of human E2F1 (amino acids: 69-223) expressed in E. Coli. MA5-17070 detects E2F1 which has a predicted molecular weight of approximately 46.9kDa.
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F2 and E2F3, have an additional cyclin binding domain. This protein binds preferentially to retinoblastoma protein pRB in a cell-cycle dependent manner. It can mediate both cell proliferation and p53-dependent/independent apoptosis.
Specifications
| E2F1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q01094, Q61501 | |
| E2F1 | |
| Purified recombinant fragment of human E2F1 (amino acids: 69-223) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 8G9 | |
| Unconjugated | |
| E2F1 | |
| E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 13555, 1869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.