Learn More
E2F-4 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98787PCP
This item is not returnable.
View return policy
Description
E2F-4 Polyclonal antibody specifically detects E2F-4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| E2F transcription factor 4, p107/p130-binding, E2F-4p107/p130-binding protein, transcription factor E2F4 | |
| Rabbit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human E2F-4. | |
| Purified |