missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
E2F-2 Monoclonal antibody specifically detects E2F-2 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | E2F-2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1H7D6 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | E2F transcription factor 2, E2F-2transcription factor E2F2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 303-395 of human E2F-2 (Q14209). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?