missing translation for 'onlineSavingsMsg'
Learn More

E2F-2 Rabbit anti-Human, Mouse, Clone: 1H7D6, Novus Biologicals™

Product Code. 18390025 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390025 100 μg 100µL
18399686 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18390025 Supplier Novus Biologicals Supplier No. NBP316165100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

E2F-2 Monoclonal antibody specifically detects E2F-2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F-2
Applications Western Blot
Classification Monoclonal
Clone 1H7D6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias E2F transcription factor 2, E2F-2transcription factor E2F2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 303-395 of human E2F-2 (Q14209).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1870
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.