missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F-1 Antibody (1D12), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | E2F-1 |
|---|---|
| Clone | 1D12 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043235
|
Novus Biologicals
NBP3-26609-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043802
|
Novus Biologicals
NBP3-26609-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E2F-1 Monoclonal antibody specifically detects E2F-1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| E2F-1 | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1 | |
| A synthesized peptide derived from Human E2F-1 [UniProt Q01094] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1D12 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1869 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title