missing translation for 'onlineSavingsMsg'
Learn More

E260, MedChemExpress

Product Code. 16490867
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490867 50 mg 50mg
16465416 100 mg 100mg
16464568 10 mg 10mg
16443136 25 mg 25mg
16407287 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490867 Supplier Medchem Express Supplier No. HY112097
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress E260 is a Fer/FerT kinase inhibitor.

E260 is a Fer/FerT kinase inhibitor.

Specifications

Chemical Name or Material E260
CAS 1241537-79-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H34N6S
Quantity 50 mg
Solubility Information DMSO : 1 mg/mL (2.28 mM; Need ultrasonic)
SMILES CC(C)C1=CC=C(C2=CN3C(SC(N4CCC(CN5CCN(C)CC5)CC4)=N3)=N2)C=C1
Molecular Weight (g/mol) 438.63
Formula Weight 438.63
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.