Learn More
Thermo Scientific E1-ClipTip™ Electronic Single Channel Pipettes, 1250
GREENER_CHOICE
Electronic tip ejection, index finger pipetting action and ClipTip technology are ideal for repetitive pipetting tasks with microcentrifuge or test tubes.
Brand:  Thermo Scientific 4670040
Artikelnummer. 15276263
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
Electronic tip ejection, index finger pipetting action and Thermo Scientitic™ ClipTip™ technology make the Thermo Scientific™ E1-ClipTip™ Electronic Single Channel Pipettes ideal for repetitive pipetting tasks with microcentrifuge or test tubes. Feel the difference.
Innovative ClipTip interlocking tip interface technology†
  • Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette
An ideal choice for sample transfers
  • Reduce repetition and save time between labware formats with adjustable tip spacing in Equalizer models.
Unified operation for multiple users and environments
  • Personalized user interface and password protection ensure consistent operation among multiple users.
Electronic operation to help you enjoy pipetting again
  • Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
Two operating options ensure optimal function for every application
  • Choose a step based Matrix or Presets with settings for your most common applications
MyPipette™ - One click access for more efficient and unified pipetting
  • Customize your main menu view by organizing icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
Store up to 20 programs
  • Tired of continuous editing? The Programs function allows you to store 20 your most common Matrix or Presets protocols with a specific name.
A pipette for every application
  • Choose from single channels, 8- and 16-channel multichannels, or 6- and 12-channel adjustable tip spacing Equalizer multichannel pipettes within the volume range of 0.5-1250µL
Track your pipette
  • Service and calibration trackers remind you when the pipette needs to be sent to serviced
Liquid specific calibrations
  • Higher quality results with liquid specific calibrations
Li-Ion battery
  • Long lasting performance and fast recharging.
Flexible re-charging
  • Use either charging stand or a plug, you decide
2-year warranty
  • With convenient web registration
Compatible with Thermo Scientific ClipTip pipette tips.
†E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and lip tip fitting technology
TRUSTED_SUSTAINABILITY
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Zubehör anzeigen
Aktionen
Aktionen
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt