missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E/Z)-10-Hydroxydemethylnortriptyline Hydrochloride, TRC
Click to view available options
Quantity:
1 mg
25 mg
Unit Size:
1mg
25mg
Specifications
Specifications
| Chemical Name or Material | (E/Z)-10-Hydroxydemethylnortriptyline Hydrochloride |
| Molecular Formula | C18H19NO . (HCl) |
| Quantity | 1 mg |
| Synonym | 10-Hydroxydesmethylnortriptyline Hydrochloride, 5-(3-Aminopropylidene)-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-10-ol Hydrochloride |
| InChI Formula | InChI=1S/C18H19NO.ClH/c19-11-5-10-15-14-7-2-1-6-13(14)12-18(20)17-9-4-3-8-16(15)17;/h1-4,6-10,18,20H,5,11-12,19H2;1H/b15-10-; |
| SMILES | OC1C2=C(C=CC=C2)/C(C3=CC=CC=C3C1)=C\CCN.Cl |
| IUPAC Name | (Z)-5-(3-aminopropylidene)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol hydrochloride |
| Molecular Weight (g/mol) | 301.81 |
| Formula Weight | 301.61 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction