missing translation for 'onlineSavingsMsg'
Learn More

(E)-Thiothixene, TRC

Product Code. 30449571
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
Product Code. Quantity unitSize
30449571 1 mg -
30452202 10 mg -
30453038 5 mg 5mg
3 options
This item is not returnable. View return policy

Product Code. 30449571

Brand: Toronto Research Chemicals TRCT3752801MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3313-27-7
Formula Weight 443.1701
InChI Formula InChI=1S/C23H29N3O2S2/c1-24(2)30(27,28)18-10-11-23-21(17-18)19(20-7-4-5-9-22(20)29-23)8-6-12-26-15-13-25(3)14-16-26/h4-5,7-11,17H,6,12-16H2,1-3H3/b19-8+
IUPAC Name (9E)-N,N-dimethyl-9-[3-(4-methylpiperazin-1-yl)propylidene]thioxanthene-2-sulfonamide
Molecular Formula C23 H29 N3 O2 S2
Molecular Weight (g/mol) 443.63
SMILES CN(C)S(=O)(=O)c1ccc2Sc3ccccc3\C(=C/CCN4CCN(C)CC4)\c2c1
Synonym (E)-N,N-Dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]-9H-thioxanthene-2-sulfonamide, (E)-N,N-dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]-thioxanthene-2-sulfonamide, (E)-Thiothixene, P 4657A, trans-Thiothixene
Chemical Name or Material (E)-Thiothixene

Specifications

Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.