missing translation for 'onlineSavingsMsg'
Learn More

(E)-SIS3, MedChemExpress

Product Code. 16483346
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16483346

Brand: Medchem Express HY13013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (E)-SIS3 is a potent and selective inhibitor of Smad3 with an IC50 of 3 μM for Smad3 phosphorylation. (E)-SIS3 inhibits the myofibroblast differentiation of fibroblasts by TGF-β1.

(E)-SIS3 is a potent and selective inhibitor of Smad3 with an IC50 of 3 μM for Smad3 phosphorylation. (E)-SIS3 inhibits the myofibroblast differentiation of fibroblasts by TGF-β1.

Specifications

Chemical Name or Material (E)-SIS3
CAS 521984-48-5
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C28H28ClN3O3
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (255.11 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES [H]Cl.O=C(/C=C/C1=C(N(C2=NC=CC=C21)C)C3=CC=CC=C3)N4CC5=C(CC4)C=C(C(OC)=C5)OC.[E]
Molecular Weight (g/mol) 489.99
Formula Weight 489.99
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.