missing translation for 'onlineSavingsMsg'
Learn More

(E)-N'-[4-Chloro-2-(trifluoromethyl)phenyl]-2-propoxy-ethanimidamide, TRC

Product Code. 30340026
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340026 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340026 Supplier Toronto Research Chemicals Supplier No. TRCC10980010MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triflumizole Metabolite FM-6-1
CAS 109849-99-2
Purity Grade Notes HPLC
Molecular Formula C12 H14 Cl F3 N2 O
Quantity 10 mg
Synonym (E)-N'-[4-Chloro-2-(trifluoromethyl)phenyl]-2-propoxy-ethanimidamide
InChI Formula InChI=1S/C12H14ClF3N2O/c1-2-5-19-7-11(17)18-10-4-3-8(13)6-9(10)12(14,15)16/h3-4,6H,2,5,7H2,1H3,(H2,17,18)
SMILES CCCOC\C(=N\c1ccc(Cl)cc1C(F)(F)F)\N
IUPAC Name N-[4-chloro-2-(trifluoromethyl)phenyl]-2-propoxy-acetamidine
Molecular Weight (g/mol) 294.70057
Formula Weight 294.0747
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.