missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Ultra Low Range DNA Ladder

Product Code. 15746548
Change view
Click to view available options
Quantity:
100 applications
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15746548 100 applications 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15746548 Supplier Invitrogen™ Supplier No. 10488096

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

DNA Ladder and 1 vial 1X E-Gel™ Sample Loading Buffer

Ready-to use DNA Ladder for use with routine throughput precast E-Gel™ agarose gels.

Invitrogen E-Gel Ultra Low Range DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 10 bp to 300 bp. E-Gel Ultra Low Range DNA Ladder consists of nine individual chromatography-purified DNA fragments with a reference band at 100 bp for easy orientation.

E-Gel Ultra Low Range DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.

Highlights of E-Gel Ultra Low Range DNA Ladder:

  • Performance—optimized for use on 4% E-Gel agarose gels
  • Sharp, clear bands—chromatography-purified fragments for consistent and reliable results, stable at room temperature up to six months
  • Ready to use—premixed with loading buffer
  • Convenient—provided with 1X E-Gel Sample Loading Buffer
  • Precise—an exact amount of DNA in each band
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 x 1 mL E-Gel Ultra Low Range DNA Ladder
• 1.25 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods store at -20°C.
Concentration 0.025 μg/μL
Gel Type Agarose
Ready to Load Yes
Sample Loading Volume 1.25 mL
Size Range 10 to 300 bp
Volume (Metric) 2 x 1000 μL
Gel Compatibility Double Comb E-Gels, Single Comb E-Gels
Quantity 100 applications
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Type DNA Ladder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.