missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ with SYBR™ Safe Mix Pack

Product Code. 31759271
Change view
Click to view available options
Quantity:
10 Gels
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31759271 10 Gels 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31759271 Supplier Invitrogen™ Supplier No. A57342
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The E-Gel with SYBR Safe Mix Pack allows researchers to try all standard format E-Gel agarose gels that use SYBR safe as a DNA stain in your workflow.

The E-Gel with SYBR Safe Mix Pack allows researchers to try all standard format E-Gel agarose gels that use SYBR safe as a DNA stain in your workflow. SYBR Safe-stained gels minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV light. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.

Benefits of using E-Gel agarose gels with SYBR Safe include:

  • Better sample integrity—exploits blue-light excitation wavelength to minimize DNA damage, as opposed to UV light
  • Safer choice—contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
  • High performance—detect 3 ng of sample DNA with a separation time as short as 13 minutes
  • Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
  • No buffers—E-Gel agarose gels do not require buffers to run

The box contains:

  • E-Gel Agarose Gels with SYBR Safe DNA Gel Stain, 1%
  • E-Gel Agarose Gels with SYBR Safe DNA Gel Stain, 2%
  • E-Gel CloneWell II Agarose Gels with SYBR Safe, 0.8%
  • E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1%
  • E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2%

Running and imaging devices

E-Gel agarose gels with SYBR Safe require the E-Gel Power Snap or E-Gel Power Snap Plus electrophoresis systems for gel running and viewing. The systems consist of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.

TRUSTED_SUSTAINABILITY

Specifications

Format Precast Gel
Gel Percentage 0.8 to 4%
Gel Type E-Gel
Separation of DNA
Separation Type Electrophoresis
Wells 11-well
Quantity 10 Gels
Sterility Non-sterile

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.