missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Sizing DNA Ladder
GREENER_CHOICE

Product Code. 15756548
Change view
Click to view available options
Quantity:
100 applications
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15756548 100 applications 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15756548 Supplier Invitrogen™ Supplier No. 10488100
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Includes

DNA Ladder and 1 vial 1X E-Gel™ Sample Loading Buffer

Ready-to use DNA Ladder for use with routine throughput precast E-Gel™ agarose gels.

Invitrogen E-Gel Sizing DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 50 to 1,500 bp. E-Gel Sizing DNA Ladder consists of 19 individual chromatography-purified DNA fragments with reference bands at 1000, 500, and 200 bp for easy orientation.

E-Gel Sizing DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.

Highlights of E-Gel Sizing DNA Ladder:

  • Performance—optimized for use on E-Gel SizeSelect ll agarose gels
  • Sharp, clear bands—chromatography-purified fragments for consistent and reliable results, stable at room temperature for up to six months
  • Ready to use—premixed with loading buffer
  • Convenient—provided with 1X E-Gel Sample Loading Buffer
  • Precise—an exact amount of DNA in each band
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 x 1.25 mL E-Gel Sizing DNA Ladder
• 1.25 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods store at -20°C.
Concentration 0.002 μg/μL
Gel Type Agarose
Ready to Load Yes
Sample Loading Volume 1.25 mL
Size Range 50 to 1500 bp
Volume (Metric) 2 x 1.25 mL
Gel Compatibility Single Comb E-Gels, SizeSelect™ II E-Gels
Quantity 100 applications
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Type DNA Ladder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.