missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. 10488100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10488100 100 g Glass bottle 100g
10366690 500 g Glass bottle 500g
10076253 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10488100 Supplier Thermo Scientific Acros Supplier No. 147431000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-70-9 | Molecular Formula: C3H6Cl2Si | Molecular Weight: 141.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-70-9
MDL Number MFCD00000492
InChI Key YLJJAVFOBDSYAN-UHFFFAOYSA-N
Synonym dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane
PubChem CID 31299
IUPAC Name dichloro-ethenyl-methylsilane
SMILES C[Si](C=C)(Cl)Cl

Specifications

Color Colorless to Brown
Density 1.0800g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H6Cl2Si
Linear Formula H2C=CHSi(CH3)Cl2
Refractive Index 1.4280 to 1.4310 (20°C, 589nm)
Quantity 100 g
Molecular Weight (g/mol) 141.066
Formula Weight 141.07
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Dichloromethylvinylsilane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.