missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Simple Runner Electrophoresis Device

Product Code. 15706558
Click to view available options
For Use With (Equipment):
SureCast Vertical Gel System
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15706558

Brand: Invitrogen™ G8000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

E-Gel Simple Runner Device,power cable with universal adapter

Easy-to-use automated device designed for use with routine throughput precast E-Gel™ agarose gels.

The Invitrogen E-Gel Simple Runner Electrophoresis Device is an easy-to-use automated device designed for use with routine-throughput E-Gel precast agarose gels. The E-Gel Simple Runner device comes pre-programmed with 15 minute and 30-minute protocols for simple and convenient device operation.

Features include:

  • Fast DNA separation—sample separation in minutes
  • Convenient—small footprint and lightweight
  • Easy to use—intuitive interface and pre-programmed protocols

Unsurpassed convenience of dry precast E-Gel agarose gels
Run DNA samples in 15-30 minutes with dry, precast, pre-stained E-Gel agarose gels. The E-Gel dry precast agarose gel technology provides speed and convenience by eliminating any need for gel preparation or staining. E-Gel agarose gels are available in a variety of gel percentages and a choice of two DNA stains.

The E-Gel Simple Runner device is compatible with E-Gel single- and double-comb agarose gels with ethidium bromide or SYBR Safe stains.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 electrophoresis device
• 1 universal power adapter

For Use With (Equipment) SureCast Vertical Gel System
Product Line E-Gel
Type Simple Runner Electrophoresis Device
Quantity 1 device
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.