missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ iBase™ Power System (EU only)
E-Gel™ iBase™ Power System (EU only)
Brand: Invitrogen™ G6400EU
Code : NEW
Additional Details : Weight : 0.50000kg
Description
The E-Gel™ iBase™ Power System gives you all the convenience of the original E-Gel™ Power Base with even more power and control. You’ll get:The E-Gel™ Safe Imager™ Real-Time Transilluminator allows you to watch migration of DNA in real-time in E-Gels containing SYBR™ Safe. It offers:
Self-contained Power System
The E-Gel™ iBase™ Power System is compatible with all low throughput E-Gels. This self-contained device includes a built-in power supply. Simply connect the AC adaptor provided and plug into an electrical outlet. View program selection and running time on the easy-to-read LCD display. Preset programs are available for various gel types or you can manually set your own run times. A reverse program allows you to change the field direction, ensuring complete capture of DNA when using the E-Gel™ CloneWell™ gels. The Speed E-Gel™ pre-set program allows separation of DNA in 7 minutes over a 2.5cm run length, and can be used with 0.8%, 1.2% or 2% E-Gels. In addition, the E-Gel™ iBase™ Power System includes an automatic shutoff feature, so you won’t overrun your gel. When running E-Gels containing SYBR™ Safe, you can safely watch the DNA bands migrate through the gel by using the E-Gel™ Safe Imager™ Real-Time Transilluminator together with the E-Gel™ iBase™ Power System. The use of SYBR™ Safe DNA Gel Stain and a Safe Imager™ Transilluminator ensures maximum possible safety to the user and gives improved cloning efficiency over ethidium bromide and UV.
Real-Time Transilluminator Designed for Safety and Cloning
The E-Gel™ Safe Imager™ Real-Time Transilluminator is designed to fit perfectly beneath the E-Gel™ iBase™ Power System and is intended for use with E-Gels containing SYBR™ Safe. Blue light emitted by the instrument is ideal for viewing DNA that is stained with SYBR™ Safe. Unlike UV, blue light causes minimal damage to DNA, offering improved safety for the user and enhanced cloning efficiency of any DNA retrieved from the gels. The E-Gel™ Safe Imager™ Real-Time Transilluminator can be purchased as a stand-alone item (Cat. No. G6500), in combination with the E-Gel™ iBase™ Power System(G6465), or as part of a Starter Kit that also includes an E-Gel™ iBase™ Power System, E-Gel™ CloneWell™ gels, and an E-Gel™ 96 High Range DNA Marker (G6500ST).
To learn more about E-Gel™ CloneWell™ gels and other E-Gels containing SYBR™ Safe, please visit www.invitrogen.com/egels
Specifications
CE, cUL US Listed, E189045, EN60825-1, Underwriters Laboratories Inc. regulation, European Community Safety, part 15 of FCC rules | |
E-Gel™ iBase™ Power System (EU only), 18.4 x 11 x 5.75cm, 1A, 240VAC, LCD Display, Programmable Control, Compatibility with All Low Throughput E-Gels, The E-Gel™ iBase™ Power System Gives you All the Convenience of the Original E-Gel™ Power Base with Even More Power and Control | |
E-Gel™ iBase™ Power System (EU Only) | |
41°F to 104°F | |
1.1 lb. | |
100 to 240 VAC 50/60 Hz 1.0 A | |
100 to 240 VAC | |
50/60 Hz |
1 A | |
Yes | |
LCD | |
5°C to 40°C | |
500 g | |
240 VAC | |
1 A |
For Research Use Only. Not for use in diagnostic procedures.