Learn More
Invitrogen™ E-Gel™ High-Throughput Mix Pack
Product Code. 31759250
Change view
Click to view available options
Quantity:
8 Gels
Unit Size:
1 set
Product Code. Quantity unitSize
31759250 8 Gels 1 set
1 options
Product Code. 31759250 Supplier Invitrogen™ Supplier No. A57344
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
The E-Gel High-Throughput Mix Pack allows researchers to try all of our high-throughput format E-Gel agarose gels.
The E-Gel High-Throughput Mix Pack allows researchers to try all of our high-throughput format E-Gel agarose gels. These gels use SYBR safe as a DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV light. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Benefits of using E-Gel agarose gels with SYBR Safe include:
  • Better sample integrity—blue-light excitation wavelength minimizes DNA damage, as opposed to UV light
  • Safer choice—contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
  • High performance—detect 3 ng of sample DNA with a separation time as short as 12 minutes
  • No buffers—E-Gel agarose gels do not require buffers to run
The box contains:
  • E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
  • E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
  • E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
  • E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
Running and imaging devices
High-throughput E-Gel agarose gels with SYBR Safe require the E-Gel Power Snap Plus Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
TRUSTED_SUSTAINABILITY
Specifications
Format Precast Gel
Gel Type E-Gel
Separation of DNA
Separation Range 100 to 2000 bp
Separation Type Electrophoresis
Quantity 8 Gels
Sterility Non-sterile
For Research Use Only.
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.