Learn More
Invitrogen™ E-Gel™ EX Mix Pack
Product Code. 31759268
Change view
Click to view available options
Quantity:
10 Gels
Unit Size:
1 set
Product Code. Quantity unitSize
31759268 10 Gels 1 set
1 options
Product Code. 31759268 Supplier Invitrogen™ Supplier No. A57343
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
The E-Gel EX Mix Pack allows researchers to try all of our standard format E-Gels EX agarose gels.
  • These pre-cast agarose gels are designed for ultrasensitive and convenient DNA sample electrophoresis in as little as eight minutes
  • Detect 1 ng of sample DNA
  • Complete DNA sample separation in eight minutes
  • Openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
  • Precast E-Gel agarose gels are ready for use and do not require gel preparation or additional buffers
  • E-Gel agarose gels do not require buffers to run
Mix Pack includes:
  • E-Gel EX Agarose Gels, 1%
  • E-Gel EX Agarose Gels, 2%
  • E-Gel EX Double Comb Agarose Gels, 1%
  • E-Gel EX Double Comb Agarose Gels, 2%
  • E-Gel SizeSelect II Agarose Gels, 2%
Running and imaging devices
  • E-Gels EX agarose gels require the E-Gel Power Snap or E-Gel Power Snap Plus electrophoresis systems for gel running and viewing
  • The systems consist of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis
TRUSTED_SUSTAINABILITY
Specifications
Format Precast Gel
Gel Type E-Gel
Separation of DNA
Separation Type Electrophoresis
Wells 11-well
Quantity 10 Gels
Sterility Non-sterile