missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ 96 Agarose Gels, 1% with E-Gel™ High Range Ladder
Click to view available options
Quantity:
8 Pack
Unit Size:
1 set
Description
E-Gel™ 96 gels are bufferless, pre-cast agarose gels designed for fast, high-throughput DNA electrophoresis. Each gel contains 96 sample lanes and 8 marker lanes that are ideal for screening multiple PCR products, plasmid preparations, and restriction digests. The unique 96-well, staggered-well format provides a 1.6-cm run length that resolves DNA between 100 bp and 10 kb in length. The E-Gel™ 96 loading format is compatible with multi-channel pipettors and the most commonly used 8-, 12-, and 96-pin liquid handling robots. With 12-minute run times, up to 20,000 samples can be resolved in a single day.
For your convenience, E-Gel™ 96 High Range DNA Markers are included with this item.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For your convenience, E-Gel™ 96 High Range DNA Markers are included with this item.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Specifications
| Description | E-Gel™ 96 Agarose Gels, 8 Gels, 1%, E-Gel™ Gel Type, Ethidium Bromide In-Gel Dye, 96 Wells, 100bp to 10kb Separation Range, Store at Room Temperature, For Nucleic Acid Gel Electrophoresis and Blotting |
| Concentration | 1% |
| Product Type | Agarose Gels with High Range Ladder |
| Separation Range | 100 to 10000 bp |
| For Use With (Application) | DNA and RNA purification analysis, nucleic acid gel electrophoresis and blotting, agarose gel electrophoresis |
| Kit Contents | 8 Gels, 1 tube of DNA ladder |
| Quantity | 8 Pack |
| Storage Requirements | Room temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction