missing translation for 'onlineSavingsMsg'
Learn More

(E)-Fenpyroximate, TRC

Product Code. 30399885
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399885 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399885 Supplier Toronto Research Chemicals Supplier No. TRCF249750100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-Fenpyroximate
CAS 134098-61-6
Purity Grade Notes HPLC
Molecular Formula C24 H27 N3 O4
Quantity 100 mg
Synonym Benzoic acid, 4-[[[(E)-[(1,3-dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]-, 1,1-dimethylethyl ester, Benzoic acid, 4-[[[[(1,3-dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]-, 1,1-dimethylethyl ester, (E)-, (E)-Fenpyroximate, Acari, Acari (pesticide), Assault, Danitron, Fenpiroximate, Fenpyroximate, Fenpyroxymate, FujiMite, Kendo, Kiron, NAI 2399-2, NNI 850, Naja, Naja (pesticide), Ortus, Ortus 05SC, Portal, tert-Butyl 4-([[(E)-1,3-dimethyl-5-phenoxypyrazol-4-yl]methyl]aminooxymethyl)benzoate
InChI Formula InChI=1S/C24H27N3O4/c1-17-21(22(27(5)26-17)30-20-9-7-6-8-10-20)15-25-29-16-18-11-13-19(14-12-18)23(28)31-24(2,3)4/h6-15H,16H2,1-5H3/b25-15+
SMILES Cc1nn(C)c(Oc2ccccc2)c1\C=N\OCc3ccc(cc3)C(=O)OC(C)(C)C
IUPAC Name tert-butyl 4-[[(E)-(1,3-dimethyl-5-phenoxypyrazol-4-yl)methylideneamino]oxymethyl]benzoate
Molecular Weight (g/mol) 421.49
Formula Weight 421.2002
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.