missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-Fenpyroximate, TRC
Specifications
Specifications
| Chemical Name or Material | (E)-Fenpyroximate |
| CAS | 134098-61-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H27 N3 O4 |
| Quantity | 250 mg |
| Synonym | Benzoic acid, 4-[[[(E)-[(1,3-dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]-, 1,1-dimethylethyl ester, Benzoic acid, 4-[[[[(1,3-dimethyl-5-phenoxy-1H-pyrazol-4-yl)methylene]amino]oxy]methyl]-, 1,1-dimethylethyl ester, (E)-, (E)-Fenpyroximate, Acari, Acari (pesticide), Assault, Danitron, Fenpiroximate, Fenpyroximate, Fenpyroxymate, FujiMite, Kendo, Kiron, NAI 2399-2, NNI 850, Naja, Naja (pesticide), Ortus, Ortus 05SC, Portal, tert-Butyl 4-([[(E)-1,3-dimethyl-5-phenoxypyrazol-4-yl]methyl]aminooxymethyl)benzoate |
| InChI Formula | InChI=1S/C24H27N3O4/c1-17-21(22(27(5)26-17)30-20-9-7-6-8-10-20)15-25-29-16-18-11-13-19(14-12-18)23(28)31-24(2,3)4/h6-15H,16H2,1-5H3/b25-15+ |
| SMILES | Cc1nn(C)c(Oc2ccccc2)c1\C=N\OCc3ccc(cc3)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl 4-[[(E)-(1,3-dimethyl-5-phenoxypyrazol-4-yl)methylideneamino]oxymethyl]benzoate |
| Molecular Weight (g/mol) | 421.49 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?