missing translation for 'onlineSavingsMsg'
Learn More

(E,E)-Piperic Acid, TRC

Product Code. 30417120
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417120 50 mg
30410466 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417120 Supplier Toronto Research Chemicals Supplier No. TRCP48153050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-72-1
Molecular Formula C12 H10 O4
Molecular Weight (g/mol) 218.21
IUPAC Name (2E,4E)-5-(1,3-benzodioxol-5-yl)penta-2,4-dienoic acid
SMILES OC(=O)\C=C\C=C\c1ccc2OCOc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H10O4/c13-12(14)4-2-1-3-9-5-6-10-11(7-9)16-8-15-10/h1-7H,8H2,(H,13,14)/b3-1+,4-2+
Formula Weight 218.0579
Chemical Name or Material (E,E)-Piperic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.