missing translation for 'onlineSavingsMsg'
Learn More

E,E-Farnesylacetone, TRC

Product Code. 30402649
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402649 25 g
30401858 1 g
30403432 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402649 Supplier Toronto Research Chemicals Supplier No. TRCF10244525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1117-52-8
Molecular Formula C18 H30 O
Molecular Weight (g/mol) 262.43
Synonym Teprenone Impurity 16
IUPAC Name (5E,9E)-6,10,14-trimethylpentadeca-5,9,13-trien-2-one
SMILES CC(=CCC\C(=C\CC\C(=C\CCC(=O)C)\C)\C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C18H30O/c1-15(2)9-6-10-16(3)11-7-12-17(4)13-8-14-18(5)19/h9,11,13H,6-8,10,12,14H2,1-5H3/b16-11+,17-13+
Formula Weight 262.2297
Chemical Name or Material E,E-Farnesylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.