missing translation for 'onlineSavingsMsg'
Learn More

(E,E)-Farnesyl MTS, TRC

Product Code. 30394417
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394417 25 mg 25mg
30394023 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394417 Supplier Toronto Research Chemicals Supplier No. TRCF10246025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E,E)-Farnesyl MTS
Analyte or Component Names (E,E)-Farnesyl MTS
Shipping Condition Room Temperature
Molecular Formula C16 H28 O2 S2
Synonym (E,E)-Farnesyl Methanethiosulfonate
InChI Formula InChI=1S/C16H28O2S2/c1-14(2)8-6-9-15(3)10-7-11-16(4)12-13-19-20(5,17)18/h8,10,12H,6-7,9,11,13H2,1-5H3/b15-10+,16-12+
Solution Type Neat
SMILES CC(=CCC\C(=C\CC\C(=C\CSS(=O)(=O)C)\C)\C)C
IUPAC Name (2E,6E)-3,7,11-trimethyl-1-methylsulfonylsulfanyldodeca-2,6,10-triene
Industry Type Pharmaceutical
Molecular Weight (g/mol) 316.522
Quantity 25 mg
Formula Weight 316.153
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.