missing translation for 'onlineSavingsMsg'
Learn More

(E,E)-Bisdemethoxycurcumin, MedChemExpress

Product Code. 16458818
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458818 10 mg 10mg
16428956 100 mg 100mg
16414727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16458818 Supplier Medchem Express Supplier No. HYN0007
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bisdemethoxycurcumin(Curcumin III; Didemethoxycurcumin) is a natural derivative of curcumin with anti-inflammatory and anti-cancer activities.

Bisdemethoxycurcumin(Curcumin III

Specifications

Chemical Name or Material (E,E)-Bisdemethoxycurcumin
CAS 33171-05-0
Color Orange
Purity Grade Notes Research
Molecular Formula C19H16O4
Quantity 10 mg
Synonym (E,E)-Curcumin III (E,E)-Didemethoxycurcumin
Solubility Information DMSO : ≥ 100 mg/mL (324.33 mM)
SMILES O=C(CC(/C=C/C1=CC=C(O)C=C1)=O)/C=C/C2=CC=C(O)C=C2.[(E)].[(E)]
Molecular Weight (g/mol) 308.33
Formula Weight 308.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.