Learn More
Invitrogen™ E. coli (strain K12/DH10B) NfuA Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5144570
Product Code. 17928992
  • £322.00 / 100µL
Description
E. coli (strain K12/DH10B) NfuA Polyclonal Antibody for Western Blot, ELISA
Involved in iron-sulfur cluster biogenesis. Binds a 4Fe-4S cluster, can transfer this cluster to apoproteins, and thereby intervenes in the maturation of FeS proteins. Could also act as a scaffoldchaperone for damaged FeS proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E. coli (strain K12/DH10B) NfuA
Polyclonal
Unconjugated
nfuA
Fe/S biogenesis protein NfuA nfuA ECDH10B_3589
Rabbit
Affinity chromatography
RUO
58390684, 82263228
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
2.82 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
B1X760
nfuA
Recombinant E.coli nfuA protein (31-128aa).
100 μL
Primary
Bacteria