missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. coli serotype O157 Monoclonal Antibody (A38D), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA518199
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Purity is >90%. Recommended pairs for ELISA: MA5-18199 (capture) and MA5-18196 (conjugated, detection).
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.Specifications
E. coli serotype O157 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
A38D | |
Unconjugated | |
Escherichia coli | |
E. coli native protein | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |