missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E. coli rpsB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117501
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Belongs to the ribosomal protein S2P family.
Specifications
| E. coli rpsB | |
| Polyclonal | |
| Unconjugated | |
| rpsB | |
| rpsB | |
| Recombinant Escherichia coli 30S ribosomal protein S2 protein (2-241aa). | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0A7V0 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 58463818 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction