Learn More
Invitrogen™ E. coli J5 LPS Monoclonal Antibody (2D7/1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183152
Description
This antibody cross-reacts with K. pneumoniae and S. sonnei and S. typhimurium. Mouse anti Escherichia coli J5 LPS antibody, clone 2D7/1 recognizes E. coli J5 LPS. The J5 mutant of E. coli lacks the enzyme uridine diphosphate glucose 4-epimerase and therefore produces an incomplete LPS, deficient in galactose and all the sugars distal to the central polymers.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications
| E. coli J5 LPS | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with <0.1% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 2D7/1 | |
| Unconjugated | |
| Escherichia coli | |
| E. coli J5 cells. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.