missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. Coli 015 Antibody (15/443), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64541R
This item is not returnable.
View return policy
Description
E. Coli 015 Monoclonal antibody specifically detects E. Coli 015 in Bacteria samples. It is validated for ELISA, Immunofluorescence
Specifications
| E. Coli 015 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA, Immunofluorescence | |
| 15/443 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Escherichia coli | |
| Native, NCTC 9015 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction