missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin, p Ser838, p Ser840 Antibody (S03-3B3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19826-100ul
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| E-Cadherin | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| A synthetic phosphopeptide corresponding to residues surrounding Ser838/Ser840 of human E-Cadherin (Uniprot # P12830) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 999 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S03-3B3 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction