missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-cadherin Monoclonal Antibody (EP700Y), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514458
This item is not returnable.
View return policy
Description
MA5-14458 targets E-Cadherin in IHC (P) applications and shows reactivity with Human samples. The MA5-14458 immunogen is a synthetic peptide derived from the fifth cadherin domain of human cadherin.
E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.
Specifications
| E-cadherin | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 0.09% sodium azide | |
| P12830 | |
| CDH1 | |
| A synthetic peptide derived from the fifth cadherin domain of human cadherin | |
| RUO | |
| 999 | |
| 4° C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| EP700Y | |
| Unconjugated | |
| CDH1 | |
| AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction