missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin Antibody (DECMA-1), Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB120-11512
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Human, Mouse, Bovine, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Microarray.
Specifications
| E-Cadherin | |
| Monoclonal | |
| Unconjugated | |
| Raw ascites with 15mM Sodium Azide | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| Rat | |
| Unpurified | |
| RUO | |
| Primary | |
| Antibody localizes the cell surface glycoprotein uvomorulin/E-cadherin | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| DECMA-1 | |
| Western Blot 1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1600, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Microarray | |
| P09803 | |
| CDH1 | |
| Mouse embryonal carcinoma cell line PCC4 Aza RI. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| 999 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction