missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin Antibody (CDH1/1525), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | E-Cadherin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CDH1/1525 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin |
| Host Species | Mouse |
| Immunogen | Recombinant human E-Cadherin protein (Uniprot: P12830) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?