Learn More
3M™ E-A-R™ Flexicap™ Replacement Pods
Click to view available options
:
Color: Yellow
Description
- Replacement foam ear pods for the E-A-RCaps™, E-A-RBand™ and Flexicaps™ Banded Earplugs
- Easy to remove and replace the pods on your banded earplugs
- Replacement pods are an easy way to improve hygiene and comfort
Specifications
Specifications
| Color | Yellow |
| Certifications/Compliance | CE Approved |
| Pairs per Package | 50 Pairs/Box |
| Noise Reduction Rating (NRR) | 23 dB |
| Dielectric | No |