Learn More
3M™ E-A-R™ Classic™ Earplugs PB-01-000
Product Code. 15749897 Shop All 3M Products
Change view
Click to view available options
Color: Yellow
Product Code.
15749897 Color: Yellow
1 options
Product Code. 15749897 Supplier 3M™ Supplier No. 7000104116
  • ONLINE EXCLUSIVE PRICE
    £61.00
    £34.48 / 200 pairs
    Save £26.52
    43% Off
View all promotions
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
3M™ E-A-R™ Classic™ Earplugs PB-01-000 meet environmental needs with its proprietary foam and preferred cylindrical shape.
  • Set the standard in hearing protection as the world's first foam earplugs
  • Firm material with slow recovery time helps achieve good fit
  • Moisture-resistant material helps maintain a consistent expansion time, even in humid environments
  • Low equilibrium pressure helps reduce pressure in ear canal thus increasing comfort and wearability
  • Exposed cell surface texture resists movement and helps maintain effective seal
  • Excellent sound attenuation characteristics
  • Supplied in re-sealable pillow-pack for ease of use
  • Sweat and humidity resistant which helps improve comfort- ideally suited for use in hot and humid conditions
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Certifications/Compliance CE Approved
Packaging Type Polybag
Pairs per Package 200 Pairs/Bag
Noise Reduction Rating (NRR) 28 dB
Corded Uncorded
Product Type Ear Plugs
Dielectric No
Material PVC