missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-9-(Prop-1-enyl)-9H-purin-6-amine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(E)-9-(Prop-1-enyl)-9H-purin-6-amine
Specifications
Specifications
| Chemical Name or Material | (E)-9-(Prop-1-enyl)-9H-purin-6-amine |
| CAS | 1446486-33-4 |
| Analyte or Component Names | (E)-9-(Prop-1-enyl)-9H-purin-6-amine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H9 N5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 9-(1E)-1-Propen-1-yl-9H-purin-6-amine, Tenofovir Disoproxil Related Compound B |
| InChI Formula | InChI=1S/C8H9N5/c1-2-3-13-5-12-6-7(9)10-4-11-8(6)13/h2-5H,1H3,(H2,9,10,11)/b3-2+ |
| SMILES | C\C=C\n1cnc2c(N)ncnc12 |
| IUPAC Name | 9-[(E)-prop-1-enyl]purin-6-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction