missing translation for 'onlineSavingsMsg'
Learn More

E 64, TRC

Product Code. 30387156
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387156 100 mg -
30381269 50 mg 50mg
30375933 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387156 Supplier Toronto Research Chemicals Supplier No. TRCE100490100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66701-25-5
Molecular Formula C15 H27 N5 O5
Molecular Weight (g/mol) 357.41
Synonym (2S,3S)-3-[[[(1S)-1-[[[4-[(Aminoiminomethyl)amino]butyl]amino]carbonyl]-3-methylbutyl]amino]carbonyl]oxiranecarboxylic Acid, [2S-[2α,3β(R*)]]-3-[[[1-[[[4-[(Aminoiminomethyl)amino]butyl]amino]carbonyl]-3-methylbutyl]amino]carbonyl]oxiranecarboxylic Acid, Proteinase Inhibitor E 64, trans-Epoxysuccinyl-L-leucylamido-(4-guanidino)Butane
IUPAC Name (2R,3R)-3-[[(2S)-1-(4-carbamimidamidobutylamino)-4-methyl-1-oxopentan-2-yl]carbamoyl]oxirane-2-carboxylic acid
SMILES CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(=O)O)C(=O)NCCCCNC(=N)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H27N5O5/c1-8(2)7-9(20-13(22)10-11(25-10)14(23)24)12(21)18-5-3-4-6-19-15(16)17/h8-11H,3-7H2,1-2H3,(H,18,21)(H,20,22)(H,23,24)(H4,16,17,19)/t9-,10+,11+/m0/s1
Formula Weight 357.2012
Chemical Name or Material E 64
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.